missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Superose™ Media Prep Grade Lab Packs

High performance gel filtration media for scaleup to preparative purifications of biomolecules

Brand:  Cytiva 17-0536-01


 View more versions of this product

Product Code. 11514925

  • £672.00 / 125mL
Estimated Shipment: 22-10-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Permit high recovery of a wide variety of biomolecules—proteins, peptides, polynucleotides, etc.
  • Supplied as bulk filtration media, for preparative separations
  • Intended as a complement to Superose PC prepacked columns; can be efficiently packed under medium pressure
  • Superose 6 Prep Grade intended for biomolecules of a wide fractionation range; Superose 12, for higher-molecular weight proteins

Order Info

Supplied in 125mL containers

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Each
30 to 40 μm
≈100mL
Separations of Proteins and other biomolecules according to size
3 to 12
Proteins, Peptides, Polynucleotides and wide variety of biomolecules
40 cm/hr.
AKTA™ design pump or system
Superose Media Prep Grade Lab Packs
100psi
Highly Cross-linked Agarose
4° to 30°C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • superose 12 prep grade

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers