missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Superose™ Media Prep Grade Lab Packs
High performance gel filtration media for scaleup to preparative purifications of biomolecules
£672.00
Specifications
Quantity | Each |
---|---|
Flow Rate | 40 cm/hr. |
Particle Size | 30 to 40 μm |
For Use With (Equipment) | AKTA™ design pump or system |
Bed Volume | ≈100mL |
Product Code | Brand | Product Type | Max. Pressure | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Max. Pressure | Price | Quantity & Availability | ||||
11544915
|
Cytiva
17-0489-01 |
Media Prep Grade Lab Packs | 58psi |
£672.00
125mL |
Estimated Shipment: 22-10-2024
Log in to see stock. |
||||
11514925
|
Cytiva
17-0536-01 |
Superose Media Prep Grade Lab Packs | 100psi |
£672.00
125mL |
Estimated Shipment: 22-10-2024
Log in to see stock. |
||||
Description
- Permit high recovery of a wide variety of biomolecules—proteins, peptides, polynucleotides, etc.
- Supplied as bulk filtration media, for preparative separations
- Intended as a complement to Superose PC prepacked columns; can be efficiently packed under medium pressure
- Superose 6 Prep Grade intended for biomolecules of a wide fractionation range; Superose 12, for higher-molecular weight proteins
Order Info
Supplied in 125mL containers
Specifications
Each | |
30 to 40 μm | |
≈100mL | |
3 to 12 | |
Proteins, Peptides, Polynucleotides and wide variety of biomolecules |
40 cm/hr. | |
AKTA™ design pump or system | |
Separations of Proteins and other biomolecules according to size | |
Highly Cross-linked Agarose |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Superose 6 prep grade, 125 ml
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.