missing translation for 'onlineSavingsMsg'
Learn More
Suramin hexasodium salt, 98%, Thermo Scientific Chemicals Product Code.: 10326401

Suramin hexasodium salt, 98%, Thermo Scientific Chemicals

Product Code. 10326401
50 mg, Glass bottle
Click to view available options
Quantity:
50 mg
250 mg
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10326401

Brand: Thermo Scientific Acros 328540500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129-46-4
Molecular Formula C51H34N6Na6O23S6
Molecular Weight (g/mol) 1429.15
MDL Number MFCD00210217
InChI Key VAPNKLKDKUDFHK-UHFFFAOYSA-H
Synonym suramin sodium, suramin hexasodium, suramin hexasodium salt, suramin sodium salt, antrypol, moranyl, naganinum, naphuride sodium, fourneau 309, suramine sodium saltShow MoreShow Less
PubChem CID 8514
IUPAC Name hexasodium;8-[[4-methyl-3-[[3-[[3-[[2-methyl-5-[(4,6,8-trisulfonatonaphthalen-1-yl)carbamoyl]phenyl]carbamoyl]phenyl]carbamoylamino]benzoyl]amino]benzoyl]amino]naphthalene-1,3,5-trisulfonateShow MoreShow Less
SMILES [Na+].[Na+].[Na+].[Na+].[Na+].[Na+].CC1=CC=C(C=C1NC(=O)C1=CC(NC(=O)NC2=CC(=CC=C2)C(=O)NC2=CC(=CC=C2C)C(=O)NC2=C3C(C=C(C=C3S([O-])(=O)=O)S([O-])(=O)=O)=C(C=C2)S([O-])(=O)=O)=CC=C1)C(=O)NC1=C2C(C=C(C=C2S([O-])(=O)=O)S([O-])(=O)=O)=C(C=C1)S([O-])(=O)=OShow MoreShow Less

Specifications

Color White
Elemental Analysis 97.5% min.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 50 mg
Merck Index 15, 9135
Solubility Information Solubility in water: soluble in water
Formula Weight 1429.1
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Suramin hexasodium salt
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0450909 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number328540500,328542500
Lot NumberA0394953 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number328540500,328542500
Lot NumberA0412708 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number328540500,328542500
Lot NumberA0417341 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number328540500,328542500
Lot NumberA0424093 Certificate TypeCertificate of Analysis Date04/11/2020 Catalog Number328540500,328542500
Product Identifier
  • Suramin hexasodium salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Suramin hexasodium salt, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.