Learn More
Poulten Selfe™ Suspended-Level Viscometers
Suitable for transparent liquids. With UKAS certificate of calibration.
Brand: Poulten Selfe™ 1625/07
This item is not returnable.
View return policy
Description
- Type BS/IP/SL, IP71.Suitable for transparent liquids. Nominal overall length 330mm. Approximate sample size 22mL to 40mL. Available without certificate of calibration, please enquire.
- With UKAS certificate of calibration.
Specifications
Suspended-Level Viscometer | |
UKAS | |
18 mL |
2.000 to 10.000 cSt | |
4 |
Product Content Correction
Product Title
Poulten Selfe™ Suspended-Level Viscometers > Size: 3A; Viscosity Range: 600 to 3000mm2/s
Spot an opportunity for improvement?Share a Content Correction