Learn More
Agilent BioTek Synergy Neo2 Hybrid Multi-Mode Microplate Reader
![GREENER_CHOICE](/content/dam/fishersci/glyphs/green_glyph.png)
BioTek Synergy Neo2 multi-mode microplate reader features patented Hybrid Technology, with independent optical paths that ensure excellent performance in all detection modes. For Research Use Only. Not for use in diagnostic procedures.
Brand: Agilent BioTek N2MBT-SN
Description
BioTek Synergy Neo2 multi-mode microplate reader features patented Hybrid Technology, with independent optical paths that ensure excellent performance in all detection modes. Variable bandwidth quad monochromators, sensitive high transmission filter-based optics, laser TRF, and up to 4 PMTs provide ultrafast measurements.
Advanced environmental controls, including CO2/O2 control, incubation to 70°C, and variable shaking support live cell assays. BioStack Neo delivers walk-away automation, high throughput, barcode-labeled filter cubes to streamline workflows and limit errors. Powerful Gen5 software provides complete reader control, powerful data analysis, and LIMS integration.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
230 to 999 nm, 1 nm increments | |
CE and TUV marked. RoHS compliant. | |
52.5 cm | |
±0.2°C at 37°C | |
0 to 4 OD | |
HTS, biomarker, nucleic acid, protein quantification, rapid kinetics, HTRF, SNP, TR FRET, BRET, Cytokines, ELISA, FP | |
41 cm | |
NEO2MBT | |
250 W max. | |
78 lb. | |
15.4 in. | |
Photodiode, Fluorescence/Alpha: Dual PMT or Single PMT (option) | |
Xenon Flash |
Monochromator: ±2 nm; <1% at 2.0 OD, <3% at 3.0 OD | |
0 to 20% | |
20.7 in. | |
Linear, Orbital, Double Orbital | |
5 to 1000 μL, 1 μL increments | |
16.1 in. | |
Monochromater: ABS, FI and LUM; Filter: Dual filter top FI/FP/TRF and LUM, Filter bottom FI/FP/TRF, TRF Laser | |
Multi-Mode Microplate Reader | |
0.0001 OD | |
35 kg | |
39 cm | |
Gen5 Data Analysis Software, Gen5 Secure for 21 CFR Part 11 compliance (option) |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.