Learn More
Sartorius Tacta™ Mechanical Pipettes
GREENER_CHOICEBIOPHARMA
Exceptionally balanced design is effortless to use and exceptionally comfortable to hold
Brand:  Sartorius LH-729230
869.99 GBP valid until 2025-04-13
Use promo code "24122" to get your promotional price.
 View more versions of this product
Product Code. 15586314
  • £1485.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Comfortable to hold due to the ergonomically designed handle
  • Unique Sartorius Optilock system provides flexibility for volume adjustment and locking
  • Controlled and smooth tip ejection with the new Sartorius Optiject technology
  • Calibration adjustment to provide accurate results for various liquid types
  • Easy to clean, with only three parts to disassemble
  • Reliable results, even over lengthy pipetting periods
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Optifit 200 μL, 350 μL, SafetySpace Filter Tips 120 μL
Autoclavable
Reusable
Polypropylene
Tacta
Non-sterile
5 to 100 μL
Yes
Yellow
Ergonomic
12
No
Pipette
Yellow
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Sartorius Tacta™ Mechanical Pipettes > Channels: 12; Color code: Yellow; Volume range: 5 - 100μL