Learn More
Thermo Scientific™ Tango™ SSTR1-bla U2OS Cells
The Tango™ SSTR1-bla U2OS cells contain the human Somatostatin Receptor 1 (SSTR1) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line.
Brand: Thermo Scientific™ K1570
Description
The Tango™ SSTR1-bla U2OS cells contain the human Somatostatin Receptor 1 (SSTR1) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line. This parental cell line stably expresses a beta-arrestin/TEV protease fusion protein and the beta-lactamase (bla) reporter gene under the control of a UAS response element.
The Tango™ SSTR1-bla U2OS cells have been functionally validated for Z' factor and EC50 concentrations of established ligands. In addition, Tango™ SSTR1-bla U2OS cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary for each product.
Specifications
NM_001049.2 | |
Dividing Cells | |
Tail from AVPR2 GPCR | |
GPCRs | |
Tango™ SSTR1-bla U2OS cells, Tango™ SSTR1-bla U2OS Assay Protocol, Certificate of Analysis | |
GeneBLAzer™ | |
8 x 106 Cells | |
BLA (Beta-Lactamase) | |
Dividing Cells | |
Tango™ |
Cell-based beta-arrestin recruitment | |
LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol. Includes: • Tango™ SSTR1-bla U2OS cells • Tango™ SSTR1-bla U2OS Assay Protocol • Certificate of Analysis The Tango™ SSTR1-bla U2OS cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt. |
|
Fluorescence | |
SSTR1 | |
Single Cell Clone Origin | |
GPCR (G-Protein Coupled Receptors) | |
End Point | |
SSTR1 | |
U2OS | |
GPCR |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.