Learn More
TaqMan™ TaqMan™ MicroRNA Reverse Transcription Kit
Provides the necessary components for 200 reactions of optimal performance in TaqMan™ MicroRNA Assays.
£210.00 - £604.00
Specifications
Detection Method | Primer-probe |
---|---|
GC-Rich PCR Performance | High |
PCR Method | 2-step RT-qPCR |
Reaction Speed | Fast |
Optimal Reaction Temperature | 42°C |
Product Code | Brand | Sufficient For | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Sufficient For | Price | Quantity & Availability | |||||
10146854
|
TaqMan™
4366596 |
200 Reactions |
£210.00
200 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10268364
|
TaqMan™
4366597 |
1000 Reactions |
£604.00
1000 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
A highly specific kit that quantitates only mature miRNAs, not precursors. A sensitive kit that conserves limited samples and requires only 1 to 10 nanograms of total RNA. It is fast, simple and scalable 2-step quantitative RT-PCR assay that provides high-quality results in less than three hours.
Components of this kit are used with the RT primer provided with the TaqMan™ MicroRNA Assay to convert miRNA to cDNA.
Order Info
Shipping Conditions: Dry Ice
Specifications
Primer-probe | |
2-step RT-qPCR | |
42°C | |
MicroRNA Reverse Transcription Kit | |
Dry Ice | |
10X | |
cDNA | |
RNA |
High | |
Fast | |
TaqMan™ | |
MultiScribe™ | |
miRNA analysis | |
10 X | |
Separate Components | |
200 bp or less |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.