Learn More
TaqMan™ TaqMan™ MicroRNA Reverse Transcription Kit
Provides the necessary components for 200 reactions of optimal performance in TaqMan™ MicroRNA Assays.
Brand: TaqMan™ 4366596
173.65 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
A highly specific kit that quantitates only mature miRNAs, not precursors. A sensitive kit that conserves limited samples and requires only 1 to 10 nanograms of total RNA. It is fast, simple and scalable 2-step quantitative RT-PCR assay that provides high-quality results in less than three hours.
Components of this kit are used with the RT primer provided with the TaqMan™ MicroRNA Assay to convert miRNA to cDNA.
Order Info
Shipping Conditions: Dry Ice

Specifications
Contains 10X RT buffer, dNTP mix w/dTTp (100M total), RNase inhibitor (20U/μL), and MultiScribe™ RT enzyme (50U/μL) for 200 reactions. Store at -15 to -25°C. | |
High | |
Fast | |
42°C | |
MicroRNA Reverse Transcription Kit | |
MultiScribe™ | |
200 Reactions | |
10X | |
cDNA | |
Separate Components | |
200 bp or less |
Primer-probe | |
2-step RT-qPCR | |
7500 System, 7500 Fast System, 7900HT System, StepOne™, StepOnePlus™, ViiA™ 7 System, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, QuantStudio 6 Pro, QuantStudio 7 Pro, QuantStudio™ 12k Flex | |
TaqMan™ | |
200 reactions | |
Dry Ice | |
miRNA analysis | |
10 X | |
200 Reactions | |
RNA |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2834912 | Certificate of Analysis | 31/01/2025 | 4366596 |
2815401 | Certificate of Analysis | 29/01/2025 | 4366596 |
2794299 | Certificate of Analysis | 29/01/2025 | 4366596 |
2794319 | Certificate of Analysis | 29/01/2025 | 4366596 |
2815408 | Certificate of Analysis | 29/01/2025 | 4366596 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.