missing translation for 'onlineSavingsMsg'
Learn More
TC-E 5005, Tocris Bioscience™ Product Code.: 30001980

TC-E 5005, Tocris Bioscience™

Product Code. 30001980
10 mg
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Product Code. 30001980

Brand: Tocris Bioscience 4572/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective PDE10A inhibitor

Potent and selective PDE10A inhibitor (IC50 values are 7.28, 239, 779, 919, 3100 and 3700 nM at PDE10A, 2A, 11A, 5A, 7B and 3A respectively and >5000 nM at PDE1B, 4A, 6, 8A and 9A). Reverses MK 801-induced hyperactivity in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 959705-64-7
Molecular Formula C15H18N4O
Molecular Weight (g/mol) 270.336
InChI Key YNADXFWEXJTQSZ-UHFFFAOYSA-N
Synonym 2-methoxy-6,7-dimethyl-9-propylimidazo 1,5-a pyrido 3,2-e pyrazine, 2-methoxy-6,7-dimethyl-9-propyl-imidazo 1,5-a pyrido 3,2-e pyrazine, 3,4-dimethyl-8-methoxy-1-propyl-imidazo 1,5-a pyrido 3,2-e pyrazine, imidazo 1,5-a pyrido 3,2-e pyrazine,2-methoxy-6,7-dimethyl-9-propyl, 12-methoxy-5,7-dimethyl-3-propyl-2,4,8,13-tetraazatricyclo 7.4.0.0 2 ,? trideca-1 13 ,3,5,7,9,11-hexaeneShow MoreShow Less
PubChem CID 24802717
SMILES CCCC1=NC(=C2N1C3=C(C=CC(=N3)OC)N=C2C)C

Specifications

Quantity 10 mg
Formula Weight 270.33
Percent Purity >99%
Chemical Name or Material TC-E 5005
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
TC-E 5005, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.