Learn More
Thermo Scientific™ Temperature Chart Recorder Paper
Use Thermo Scientific™ Temperature Chart Recorder Paper in seven-day circular temperature chart recorders.
Brand:  Thermo Scientific™ 7289
 View more versions of this product
Product Code. 10354557
  • £92.39 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Use Thermo Scientific™ Temperature Chart Recorder Paper in seven-day circular temperature chart recorders.
  • 6″ (15.2cm) diameter
  • For use in freestanding and panel-mounted chart recorders
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Chart Recorder Paper
Freestanding
Recorder
6 in.
-200°C to 0°C
Free-Standing and Panel-Mounted Chart Recorders
7 Days
15.2 cm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Temperature Chart Recorder Paper > -200°C to 0°C