Learn More
tert-Butyl methyl ether, 99.9%, extra pure, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | -110.0°C |
Density | 0.7404g/mL |
Boiling Point | 54.0°C to 56.0°C |
Quantity | 2.5 L |
Flash Point | −28°C |
Packaging | Glass Bottle |
Physical Form | Liquid |
Refractive Index | 1.3675 to 1.3695 |
Assay Percent Range | 99.9% |
Residue after Evaporation | 1mg/100mL max. |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0466034 | Certificate of Analysis | 10/09/2024 | 378720100,378720025,378720010 |
A0457574 | Certificate of Analysis | 27/11/2023 | 378720100,378720025,378720010 |
A0457575 | Certificate of Analysis | 17/11/2023 | 378720100,378720025,378720010 |
A0457573 | Certificate of Analysis | 14/11/2023 | 378720100,378720025,378720010 |
A0454726 | Certificate of Analysis | 25/08/2023 | 378720100,378720025,378720010 |
Safety and Handling


- tert-Butyl methyl ether
- Danger
- Skin corrosion/irritation Category 2
- Flammable liquid Category 2
- H315-Causes skin irritation.
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
Special Offers
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
Days
42Hours
16Minutes
50Seconds
38New Offer Save Up to 70%
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.