missing translation for 'onlineSavingsMsg'
Learn More
tert-Butyl methyl ether, HPLC grade, 99+%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals Product Code.: 11324807

tert-Butyl methyl ether, HPLC grade, 99+%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Product Code. 11324807
250 mL
Click to view available options
Quantity:
250 mL
1 L
This item is not returnable. View return policy

Product Code. 11324807

Brand: Thermo Scientific Alfa Aesar 041839.AK

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chromatography solvent

tert-Butyl methyl ether is used as a solvent for chromatography and in the manufacturing of chemicals, drugs, and drug intermediates. It is used as a mobile phase in High Performance Liquid Chromatography. It acts as a fuel component in fuel for gasoline engines. They raise the oxygen content of gasoline (oxygenates).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1634-04-4
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00008812
InChI Key BZLVMXJERCGZMT-UHFFFAOYSA-N
Synonym tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropaneShow MoreShow Less
PubChem CID 15413
ChEBI CHEBI:27642
IUPAC Name 2-methoxy-2-methylpropane
SMILES CC(C)(C)OC

Specifications

Melting Point -109°C
Density 0.742 g/mL
Boiling Point 55°C to 56°C
Quantity 250 mL
Flash Point -25°C (-13°F)
Physical Form Liquid
Refractive Index 1.369
UN Number 2350
Beilstein 1730942
Merck Index 14,6032
Solubility Information Slightly miscible with water. Highly miscible with ethanol and ethyl ether.
Formula Weight 88.15
Percent Purity ≥99%
Grade HPLC
Chemical Name or Material tert-Butyl Methyl ether
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberT12K867 Certificate TypeCertificate of Analysis Date22/10/2024 Catalog Number041839.AK,041839.K2
Lot NumberT12K868 Certificate TypeCertificate of Analysis Date06/09/2024 Catalog Number041839.AK,041839.K2
Lot NumberQ05K783 Certificate TypeCertificate of Analysis Date10/05/2024 Catalog Number041839.AK,041839.K2
Lot NumberY17J851 Certificate TypeCertificate of Analysis Date05/03/2024 Catalog Number041839.AK,041839.K2
Lot NumberQ30J702 Certificate TypeCertificate of Analysis Date11/10/2023 Catalog Number041839.AK,041839.K2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
tert-Butyl methyl ether, HPLC grade, 99+%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.