Learn More
tert-Butyl methyl ether, HPLC grade, 99+%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Description
tert-Butyl methyl ether is used as a solvent for chromatography and in the manufacturing of chemicals, drugs, and drug intermediates. It is used as a mobile phase in High Performance Liquid Chromatography. It acts as a fuel component in fuel for gasoline engines. They raise the oxygen content of gasoline (oxygenates).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under argon

Specifications
Specifications
Melting Point | -109°C |
Density | 0.742 g/mL |
Boiling Point | 55°C to 56°C |
Quantity | 1 L |
Flash Point | -25°C (-13°F) |
Odor | Mild |
Packaging | ChemSeal™ Bottle |
Physical Form | Liquid |
Refractive Index | 1.369 |
UN Number | UN2350 |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
T12K867 | Certificate of Analysis | 22/10/2024 | 041839.AK,041839.K2 |
T12K868 | Certificate of Analysis | 06/09/2024 | 041839.AK,041839.K2 |
Q05K783 | Certificate of Analysis | 10/05/2024 | 041839.AK,041839.K2 |
Y17J851 | Certificate of Analysis | 05/03/2024 | 041839.AK,041839.K2 |
Q30J702 | Certificate of Analysis | 11/10/2023 | 041839.AK,041839.K2 |
Safety and Handling


- tert-Butyl methyl ether
- Danger
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- H225-Highly flammable liquid and vapour.
- H315-Causes skin irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- MIXTURE LIST-Contains : Methyl tert-butyl ether
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.