missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butylchlorodimethylsilane, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 85.0°C to 91.0°C |
Color | Colorless to White |
Density | 0.8700g/mL |
Boiling Point | 125.0°C |
Flash Point | 22°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (GC) |
Packaging | Glass bottle |
Linear Formula | (CH3)3CSi(CH3)2Cl |
Quantity | 25 g |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0467621 | Certificate of Analysis | 30/01/2025 | 183930250,183930050,183935000,183931000 |
A0464389 | Certificate of Analysis | 08/07/2024 | 183930250,183930050,183935000,183931000 |
A0456886 | Certificate of Analysis | 11/10/2023 | 183930250,183930050,183935000,183931000 |
A0454038 | Certificate of Analysis | 25/08/2023 | 183930250,183930050,183935000,183931000 |
A0392431 | Certificate of Analysis | 12/12/2022 | 183930250,183930050,183935000,183931000 |
Safety and Handling


Product Identifier
- tert-Butylchlorodimethylsilane
- Danger
- Flammable solid Category 2
- Skin corrosion/irritation Category 1 B
- H314-Causes severe skin burns and eye damage.
- H228-Flammable solid.
- EUH014-Reacts violently with water.
- EUH029-Contact with water liberates toxic gas.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P402+P404-Store in a dry place. Store in a closed container.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
tert-Butylchlorodimethylsilane, 98%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction