missing translation for 'onlineSavingsMsg'
Learn More

tert-Butylchlorodimethylsilane, 98%, Thermo Scientific Chemicals

£32.86 - £804.00

Chemical Identifiers

CAS 18162-48-6
Molecular Formula C6H15ClSi
Molecular Weight (g/mol) 150.73
MDL Number MFCD00000501
InChI Key BCNZYOJHNLTNEZ-UHFFFAOYSA-N
Synonym tert-butyldimethylsilyl chloride, tert-butyldimethylchlorosilane, t-butyldimethylchlorosilane, tert-butylchlorodimethylsilane, tbdms chloride, tert-butyl chloro dimethylsilane, chloro-tert-butyldimethylsilane, silane, chloro 1,1-dimethylethyl dimethyl, tbscl, t-butyldimethylsilyl chloride
PubChem CID 28928
ChEBI CHEBI:85071
IUPAC Name tert-butyl-chloro-dimethylsilane
SMILES CC(C)(C)[Si](C)(C)Cl
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10164010
View Documents Promotion Details
Thermo Scientific Acros
183930050
5 g Glass Bottle
£32.86
5g
Please to purchase this item. Need a web account? Register with us today!
10275710
View Documents
Thermo Scientific Acros
183930250
25 g Glass bottle
£92.86
25g
Please to purchase this item. Need a web account? Register with us today!
10712041
View Documents
Thermo Scientific Acros
183931000
100 g Glass bottle
£277.38
100g
Please to purchase this item. Need a web account? Register with us today!
10331703
View Documents
Thermo Scientific Acros
183935000
500 g Glass bottle
£804.00
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

18162-48-6
150.73
BCNZYOJHNLTNEZ-UHFFFAOYSA-N
28928
tert-butyl-chloro-dimethylsilane
C6H15ClSi
MFCD00000501
tert-butyldimethylsilyl chloride, tert-butyldimethylchlorosilane, t-butyldimethylchlorosilane, tert-butylchlorodimethylsilane, tbdms chloride, tert-butyl chloro dimethylsilane, chloro-tert-butyldimethylsilane, silane, chloro 1,1-dimethylethyl dimethyl, tbscl, t-butyldimethylsilyl chloride
CHEBI:85071
CC(C)(C)[Si](C)(C)Cl

Specifications

18162-48-6
Colorless to White
125.0°C
Authentic
C6H15ClSi
MFCD00000501
04,176; 05,74; 06,78; 08,59; 09,77; 10,62; 13,87; 17,5
0.87
Solubility in water: reacts. Other solubilities: reacts with alcohols, organic acids and amines, soluble in ethers and hydrocarbons
CC(C)(C)[Si](C)(C)Cl
150.73
CHEBI:85071
98%
tert-Butylchlorodimethylsilane
85.0°C to 91.0°C
0.8700g/mL
22°C
97.5% min. (GC)
(CH3)3CSi(CH3)2Cl
2925
15, 1567
tert-butyldimethylsilyl chloride, tert-butyldimethylchlorosilane, t-butyldimethylchlorosilane, tert-butylchlorodimethylsilane, tbdms chloride, tert-butyl chloro dimethylsilane, chloro-tert-butyldimethylsilane, silane, chloro 1,1-dimethylethyl dimethyl, tbscl, t-butyldimethylsilyl chloride
BCNZYOJHNLTNEZ-UHFFFAOYSA-N
tert-butyl-chloro-dimethylsilane
28928
150.73
Crystals
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Flammable solid.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or ha

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 242-042-4

Product Identifier
  • tert-Butylchlorodimethylsilane

Signal Word
  • Danger

Hazard Category
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H228-Flammable solid.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.