Learn More
Retsch™ Test Sieves with 11.20 to 112.00 mm Mesh Size
Retsch™ Test Sieves consist of a solid stainless steel sieve frame with high stability, resulting in reliable sieving results.
£151.00 - £157.00
Specifications
Diameter (Metric) Outer | 200 mm |
---|---|
Type | Test Sieve |
Certifications/Compliance | DIN ISO 3310-1 |
Material | Stainless Steel |
Includes | Calibration Certificate |
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
15600409
|
Retsch™
60.131.011200 |
11.20 mm |
£151.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
15610409
|
Retsch™
60.131.012500 |
12.50 mm |
£151.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
15620409
|
Retsch™
60.131.013200 |
13.20 mm |
£151.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
15630409
|
Retsch™
60.131.014000 |
14.00 mm |
£151.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
15640409
|
Retsch™
60.131.016000 |
16.00 mm |
£151.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
15650409
|
Retsch™
60.131.018000 |
18.00 mm |
£151.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
15660409
|
Retsch™
60.131.019000 |
19.00 mm |
£151.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
15670409
|
Retsch™
60.131.020000 |
20.00 mm |
£151.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
15680409
|
Retsch™
60.131.022400 |
22.40 mm |
£151.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
15690409
|
Retsch™
60.131.025000 |
25.00 mm |
£151.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
15600419
|
Retsch™
60.131.026500 |
26.50 mm |
£151.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
15610419
|
Retsch™
60.131.028000 |
28.00 mm |
£151.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
15620419
|
Retsch™
60.131.031500 |
31.50 mm |
£157.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
15630419
|
Retsch™
60.131.035500 |
35.50 mm |
£157.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
15640419
|
Retsch™
60.131.037500 |
37.50 mm |
£157.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
15650419
|
Retsch™
60.131.040000 |
40.00 mm |
£157.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
15660419
|
Retsch™
60.131.045000 |
45.00 mm |
£157.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
15670419
|
Retsch™
60.131.063000 |
63.00 mm |
£157.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
15680419
|
Retsch™
60.131.075000 |
75.00 mm |
£157.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
15690419
|
Retsch™
60.131.080000 |
80.00 mm |
£157.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
15600429
|
Retsch™
60.131.090000 |
90.00 mm |
£157.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
15610429
|
Retsch™
60.131.100000 |
100.00 mm |
£157.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
15620429
|
Retsch™
60.131.112000 |
112.00 mm |
£157.00
Each |
Estimated Shipment: 21-10-2024
Log in to see stock. |
|||||
Description
- High quality joining technology guarantees permanently taut wire mesh
- One-piece cast sieve frame and seamless mesh joint prevent cross contamination
- Stainless steel sieve frame with high form stability
- High degree of corrosion resistance and easy cleaning thanks to high-alloy stainless steel
- Excellent product quality due to extensive optical inspection
- Maximum stability and optimum sealing when used in sieve stacks thanks to the o-ring which is placed in the recess designed for this purpose
- Clear and precise labeling of the sieves with full traceability based on individualized laser engraving
Specifications
200 mm | |
DIN ISO 3310-1 | |
Calibration Certificate |
Test Sieve | |
Stainless Steel | |
50 mm |