missing translation for 'onlineSavingsMsg'
Learn More
Tetra-n-butylammonium perchlorate, 99+%, Thermo Scientific Chemicals Product Code.: 15421177

Tetra-n-butylammonium perchlorate, 99+%, Thermo Scientific Chemicals

Product Code. 15421177
25 g
Click to view available options
Quantity:
2.5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 15421177

Brand: Thermo Scientific Alfa Aesar 030801.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Tetra-n-butylammonium perchlorate is used as a supporting electrolyte in polarographic measurements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1923-70-2
Molecular Formula C16H36ClNO4
Molecular Weight (g/mol) 341.917
MDL Number MFCD00038722
InChI Key KBLZDCFTQSIIOH-UHFFFAOYSA-M
Synonym tetrabutylammonium perchlorate, 1-butanaminium, n,n,n-tributyl-, perchlorate, tetrabutyl ammonium perchlorate, tetra-n-butylammonium perchlorate, tetrabutylammonium perchlorat, 1-butanaminium, n,n,n-tributyl-, perchlorate 1:1, acmc-1bpvk, n,n,n-tributylbutan-1-aminium perchlorate, tetrabutylammoniumperchlorate, tetrabutylazanium perchlorateShow MoreShow Less
PubChem CID 74723
IUPAC Name tetrabutylazanium;perchlorate
SMILES CCCC[N+](CCCC)(CCCC)CCCC.[O-]Cl(=O)(=O)=O

Specifications

Melting Point 210°C to 217°C
Color White
Linear Formula [CH3(CH2)3]4NClO4
Quantity 25 g
UN Number UN1481
Sensitivity Hygroscopic
Solubility Information Soluble in acetonitrile and ethanol. Slightly soluble in water.
Formula Weight 341.92
Percent Purity ≥99%
Physical Form Crystalline
Chemical Name or Material Tetra-n-butylammonium perchlorate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW27K050 Certificate TypeCertificate of Analysis Date05/10/2024 Catalog Number030801.14,030801.22,030801.88
Lot NumberP12K005 Certificate TypeCertificate of Analysis Date22/03/2024 Catalog Number030801.14,030801.22,030801.88
Lot NumberX11E028 Certificate TypeCertificate of Analysis Date19/07/2023 Catalog Number030801,030801.14,030801.22,030801.88
Lot NumberZ19I034 Certificate TypeCertificate of Analysis Date13/01/2023 Catalog Number030801.14,030801.22,030801.88
Lot NumberX18I013 Certificate TypeCertificate of Analysis Date26/10/2022 Catalog Number030801.14,030801.22,030801.88

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tetra-n-butylammonium perchlorate, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.