Learn More
Tetrabutylammonium hydroxide, 40 wt.% (1.5M) solution in water, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 176612500
98.65 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Phase transfer catalyst
Specifications
Tetrabutylammonium hydroxide | |
2052-49-5, 7732-18-5 | |
0.9950g/mL | |
Plastic bottle | |
[CH3(CH2)3]4NOH | |
250 g | |
05,645; 11,500 | |
tetrabutylammonium hydroxide, tetra-n-butylammonium hydroxide, tetrabutylazanium hydroxide, tetrabutylammoniumhydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide, ammonium, tetrabutyl-, hydroxide, n,n,n-tributyl-1-butanaminium hydroxide, tetra n-butyl ammonium hydroxide, tetra-n-butyl ammonium hydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide 1:1 | |
VDZOOKBUILJEDG-UHFFFAOYSA-M | |
tetrabutylazanium;hydroxide | |
2723671 | |
38 to 42% (Total base) |
40 wt.% Solution in Water | |
White to Yellow | |
>100.0°C | |
C16H37NO | |
MFCD00009425 | |
04, II, 634 | |
0.995 | |
Solubility in water: soluble. | |
[OH-].CCCC[N+](CCCC)(CCCC)CCCC | |
259.48 | |
259.46 | |
Crystals or Powder |
Safety and Handling
- Tetrabutylammonium hydroxide, 40 wt.% solution in water
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 A
- Acute toxicity Category 4
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0465430 | Certificate of Analysis | 14/08/2024 | 176610010,176612500,176610025,176610500 |
A0464900 | Certificate of Analysis | 25/07/2024 | 176610010,176612500,176610025,176610500 |
A0464557 | Certificate of Analysis | 12/07/2024 | 176610010,176612500,176610025,176610500 |
A0457677 | Certificate of Analysis | 14/11/2023 | 176610010,176612500,176610025,176610500 |
A0450481 | Certificate of Analysis | 25/08/2023 | 176610010,176612500,176610025,176610500 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only