missing translation for 'onlineSavingsMsg'
Learn More
Tetradecane, 99%, Thermo Scientific Chemicals Product Code.: 11934401

Tetradecane, 99%, Thermo Scientific Chemicals

Product Code. 11934401
2.5 L, Glass bottle
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
EcoSafPak™ Glass Bottle
Glass bottle
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 11934401

Brand: Thermo Scientific Acros 174130025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629-59-4
InChI Key BGHCVCJVXZWKCC-UHFFFAOYSA-N
Synonym n-tetradecane, alkanes, c14-16, tridecane, methyl, ccris 715, tetradecane, analytical standard, alkanes, c14-30, myristyl, tetradekan, n-teradecane, olefineShow MoreShow Less
PubChem CID 12389
ChEBI CHEBI:41253
IUPAC Name tetradecane
SMILES CCCCCCCCCCCCCC

Specifications

Melting Point 5.8°C
Color Colorless
Density 0.7670g/mL
Boiling Point 252.0°C to 254.0°C
Flash Point 99°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C14H30
Linear Formula CH3(CH2)12CH3
Refractive Index 1.4280 to 1.4300
Quantity 2.5 L
Beilstein 01, 171
Specific Gravity 0.767
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol
Molecular Weight (g/mol) 198.39
Formula Weight 198.39
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Tetradecane
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467953 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number174130025,174135000,174131000
Lot NumberA0455844 Certificate TypeCertificate of Analysis Date14/06/2024 Catalog Number174130025,174135000,174131000
Lot NumberA0401254 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number174130025,174135000,174131000
Lot NumberA0376073 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number174130025,174135000,174131000
Lot NumberA0441334 Certificate TypeCertificate of Analysis Date18/04/2022 Catalog Number174130025,174135000,174131000
compliance-icons
Product Identifier
  • Tetradecane
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tetradecane, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.