Learn More
TFEB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13372-1-AP-20UL
 View more versions of this product
Product Code. 16841544
  • £135.00 / 20µL
Explore more special offers
Description
Description
This is a rabbit polyclonal antibody raised against the full length of human TFEB. The calculated molecular weight of TFEB is 53 kDa, but modified TFEB is about 65-70 kDa.
The Transcription factor EB (TFEB) is a member in the basic helix-loop-helix leucine zipper superfamily of transcription factors that is translocated in a subset of renal tumors. Recent studies have shown that lysosomal biogenesis is regulated by TFEB, which is in turn regulated by the mammalian target of rapamycin (mTOR) complex 1. Other evidence suggests that TFEB coordinates the major steps of the autophagic pathway by driving the expression of autophagy and lysosomal genes.
Specifications
Specifications
TFEB
Polyclonal
Unconjugated
TFEB
AlphaTFEB, bHLHe35, TCFEB, TFEB, transcription factor EB
Rabbit
Antigen Affinity Chromatography
RUO
21425, 316214, 7942
-20°C
Liquid
Western Blot, Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry
0.35 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P19484, Q9R210
Tfeb
TFEB Fusion Protein Ag4194
20 μL
Primary
Human, Rat, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
TFEB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated