missing translation for 'onlineSavingsMsg'
Learn More
Thallium(I) chloride, 99.999% (metals basis), Thermo Scientific Chemicals Product Code.: 11371898

Thallium(I) chloride, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11371898
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11371898

Brand: Thermo Scientific Alfa Aesar 011839.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7791-12-0
Molecular Formula ClTl
Molecular Weight (g/mol) 239.83
MDL Number MFCD00011274
InChI Key GBECUEIQVRDUKB-UHFFFAOYSA-M
Synonym thallium chloride, thallium i chloride, thallous chloride, thallium monochloride, thallium 1+ chloride, tlcl, thallium chloride tlcl, thallium chloride van, rcra waste number u216, rcra waste no. u216Show MoreShow Less
PubChem CID 24642
ChEBI CHEBI:37117
IUPAC Name chlorothallium
SMILES Cl[Tl]

Specifications

Melting Point 430°C
Physical Form Powder
Quantity 5 g
UN Number UN1707
Merck Index 14,9260
Formula Weight 239.82
Percent Purity 99.999%
Refractive Index 2.247
Sensitivity Air Sensitive
Density 7.0 g/mL
Assay (metals basis)
Chemical Name or Material Thallium(I) chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberS26F009 Certificate TypeCertificate of Analysis Date14/01/2025 Catalog Number011839.22,011839.14,011839.06
Lot NumberS16A025 Certificate TypeCertificate of Analysis Date13/10/2022 Catalog Number011839.22,011839.14,011839.06
Lot NumberL07Q008 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number011839.22,011839.14,011839.06
Lot NumberJ10N29 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number011839.22,011839.14,011839.06
Lot NumberL14W004 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number011839.22,011839.14,011839.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thallium(I) chloride, 99.999% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.