missing translation for 'onlineSavingsMsg'
Learn More
Thallium(I) sulfate, 99.5% min (metals basis), Thermo Scientific Chemicals Product Code.: 11323918

Thallium(I) sulfate, 99.5% min (metals basis), Thermo Scientific Chemicals

Product Code. 11323918
250 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11323918

Brand: Thermo Scientific Alfa Aesar 040342.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Thallium(I) sulfate is used as a precursor to thallium(I) sulfide, which exhibits high electrical conductivity when exposed to infrared light. It is further used in photocells. It is also mixed with selenium and arsenic to produce low melting glasses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7446-18-6
Molecular Formula O4STl2
Molecular Weight (g/mol) 504.816
MDL Number MFCD00011278
InChI Key YTQVHRVITVLIRD-UHFFFAOYSA-L
Synonym thallous sulfate, thallium sulfate, thallium i sulfate, tharattin, zelio, bonide antzix, th-universal, unii-u9f9qir12t, rcra waste number p115, sulfuric acid, dithallium 1+ saltShow MoreShow Less
PubChem CID 24833
ChEBI CHEBI:81836
IUPAC Name thallium(1+);sulfate
SMILES [O-]S(=O)(=O)[O-].[Tl+].[Tl+]

Specifications

Boiling Point (decomposition)
Melting Point 632°C
Physical Form Crystalline
Quantity 250 g
Assay Percent Range 99.5% min (metals basis)
Formula Weight 504.8
Density 6.77
Chemical Name or Material Thallium(I) sulfate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY04H025 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number040342.A1,040342.18,040342.09,040342.30
Lot NumberP15I005 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number040342.A1,040342.18,040342.09,040342.30
Lot NumberM25I006 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number040342.A1,040342.18,040342.09,040342.30
Lot NumberM25I008 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number040342.A1,040342.18,040342.09,040342.30
Lot NumberM25I009 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number040342.A1,040342.18,040342.09,040342.30

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thallium(I) sulfate, 99.5% min (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.