missing translation for 'onlineSavingsMsg'
Learn More
Thallium(I) sulfate, Honeywell Fluka™ Product Code.: 15667370

Thallium(I) sulfate, Honeywell Fluka™

Product Code. 15667370 Shop All Honeywell Fluka Products
100 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 15667370

Brand: Honeywell-Fluka 88290100G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Purum,≥98.0% (RT)

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7446-18-6
Molecular Formula O4STl2
Molecular Weight (g/mol) 504.816
MDL Number MFCD00011278
InChI Key YTQVHRVITVLIRD-UHFFFAOYSA-L
Synonym thallous sulfate, thallium sulfate, thallium i sulfate, tharattin, zelio, bonide antzix, th-universal, unii-u9f9qir12t, rcra waste number p115, sulfuric acid, dithallium 1+ saltShow MoreShow Less
PubChem CID 24833
ChEBI CHEBI:81836
IUPAC Name thallium(1+);sulfate
SMILES [O-]S(=O)(=O)[O-].[Tl+].[Tl+]

Specifications

Quantity 100 g
Linear Formula Tl2SO4
UN Number UN1707
Formula Weight 504.83g/mol
Percent Purity ≥98% (RT)
Density 6.77g/mL at 25°C
Chemical Name or Material Thallium(I) sulfate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thallium(I) sulfate, Honeywell Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.