Learn More
Thapsigargin, 95%, Thermo Scientific Chemicals
Description
Thapsigargin- induces down regulation of the EGF receptor is independent of protein kinase C activation. It is also used as an inducer of stored Ca2+ release.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Thapsigargin- induces down regulation of the EGF receptor is independent of protein kinase C activation. It is also used as an inducer of stored Ca2+ release.
Solubility
Soluble in DMSO (20 mg/ml), ethanol (20 mg/ml), and acetone.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C.

Specifications
Specifications
Color | White |
Quantity | 1 mg |
Sensitivity | Light sensitive |
Merck Index | 14,9272 |
Synonym | thapsigargin, unii-z96bq26rzd, z96bq26rzd, chembl96926, octanoic acid 3s-3alpha, 3abeta, 4alpha, 6beta, 6abeta, 7beta, 8alpha z , 9balpha-6-acetyloxy-2,3,-3a,4,5,6,6a,7,8,9b-decahydro-3,3a-dihydroxy-3,6,9-trimethyl-8-2-methyl-1-oxo-2-butenyl oxy-2-oxo-4-1-oxobutoxy-azuleno 4,5-b furan-7-yl ester, octanoic acid, 6-acetyloxy-2,3,3a,4,5,6,6a,7,8,9b-decahydro-3,3a-dihydroxy-3,6,9-trimethyl-8-2-methyl-1-oxo-2-butenyl oxy-2-oxo-4-1-oxobutoxy azuleno 4,5-b furan-7-yl ester, 3s-3alpha,3abeta,4alpha,6beta,6abeta,7beta,8alpha z ,9balpha, tg1, --thapsigargin, octanoic acid 3s-3alpha, 3abeta, 4alpha, 6beta,6abeta, 7beta, 8alpha z , 9balpha-6-acetyloxy-2,3,-3a,4,5,6,6a,7,8,9b-decahydro-3,3a-dihydroxy-3,6,9-trimethyl-8-2-methyl-1-oxo-2-butenyl oxy-2-oxo-4-1-oxobutoxy-azuleno 4,5-b furan-7-yl ester, octanoic acid 3s-3alpha, 3abeta, 4alpha,6beta, 6abeta, 7beta, 8alpha z , 9balpha-6-acetyloxy-2,3,-3a,4,5,6,6a,7,8,9b-decahydro-3,3a-dihydroxy-3,6,9-trimethyl-8-2-methyl-1-oxo-2-butenyl oxy-2-oxo-4-1-oxobutoxy-azuleno 4,5-b furan-7-yl ester |
Solubility Information | Soluble in DMSO (20mg/ml),ethanol (20mg/ml),and acetone. |
IUPAC Name | [(3S,3aR,4S,6S,6aS,7S,8S,9bR)-6-acetyloxy-4-butanoyloxy-3,3a-dihydroxy-3,6,9-trimethyl-8-(2-methylbut-2-enoyloxy)-2-oxo-4,5,6a,7,8,9b-hexahydroazuleno[4,5-b]furan-7-yl] octanoate |
Formula Weight | 650.75 |
Percent Purity | 95% |
Physical Form | Solid Film |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
N18L045 | Certificate of Analysis | 20/02/2025 | J62866.MA,J62866.#0,J62866.M^ |
N18L038 | Certificate of Analysis | 20/02/2025 | J62866.MA,J62866.#0,J62866.M^ |
X24K036 | Certificate of Analysis | 27/01/2025 | J62866.MA,J62866.#0,J62866.M^ |
Y25K029 | Certificate of Analysis | 26/11/2024 | J62866.MA,J62866.#0,J62866.M^ |
W24K026 | Certificate of Analysis | 07/10/2024 | J62866.MA,J62866.#0,J62866.M^ |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.