Learn More
Brady™ Thermal Transfer Printable Labels
Brady™ Thermal Transfer Printable Labels are comprised of colored vinyl film available in different widths with a permanent acrylic adhesive.
£190.00 - £267.00
Specifications
Adhesive Type | Acrylic |
---|---|
Material | Pressure-sensitive Vinyl |
Length (Metric) | 40 m |
Product Code | Brand | Width (Metric) | Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Width (Metric) | Color | Price | Quantity & Availability | ||||
11508684
|
Brady™
217509 |
110 mm | White |
£267.00
1 roll |
Estimated Shipment: 15-07-2024
Log in to see stock. |
||||
11508694
|
Brady™
217521 |
63 mm | Green |
£190.00
1 roll |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||||
11528674
|
Brady™
217501 |
110 mm | Green |
£267.00
1 roll |
Estimated Shipment: 15-07-2024
Log in to see stock. |
||||
11528684
|
Brady™
217511 |
83 mm | Green |
£225.00
1 roll |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||||
11528694
|
Brady™
217523 |
63 mm | Blue |
£190.00
1 roll |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||||
11548674
|
Brady™
217503 |
110 mm | Blue |
£267.00
1 roll |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||||
11548694
|
Brady™
217525 |
63 mm | Red |
£190.00
1 roll |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||||
11558674
|
Brady™
217504 |
110 mm | Yellow |
£267.00
1 roll |
Estimated Shipment: 15-07-2024
Log in to see stock. |
||||
11558684
|
Brady™
217515 |
83 mm | Red |
£225.00
1 roll |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||||
11568674
|
Brady™
217505 |
110 mm | Red |
£267.00
1 roll |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||||
11588684
|
Brady™
217519 |
83 mm | White |
£225.00
1 roll |
Estimated Shipment: 15-07-2024
Log in to see stock. |
||||
Description
- Colored, printable vinyl film for thermal transfer printers
- Permanent acrylic adhesive
- For general and industrial identification
- Compatible with Brady printer models BBP12; BBP11; PR200Plus; PR300Plus; PR600Plus; BP1244; BP1344; BP4000; BP4320
- Supplied as 40m rolls in three widths
- Service temperatures -70°C to 40°C
Specifications
Acrylic | |
Pressure-sensitive Vinyl | |
40 m |