Learn More
Thermo Scientific™ Reagent Reservoir

Thermo Scientific™ Reagent Reservoirs are designed for use in your pipetting techniques with single and multi-channel pipettes.
Brand: Thermo Scientific™ 9510047
22.37 GBP valid until 2025-06-30
Use promo code "25076" to get your promotional price.
Description
Thermo Scientific™ Reagent Reservoirs are designed to meet all your pipetting needs. We've added features with you in mind: graduation lines guide you on remaining reagent volume, corner pour spouts prevent spilling when emptying the reservoir, and wide stable base helps to prevents tipping.
- Designed for multiple pipetting applications
- Trough within a trough design maximizes amount of liquid accessible to pipet tips when using small amounts of reagent
- Pour-off spouts in all four corners reduce spillage when pouring reagents out of reservoir Graduations on inside wall enable quick measurement of remaining liquid
- Extra-wide base adds rigidity and stability to reservoir, helping to avoid spills
- 25mL divided reservoir allows the pipetting of two different reagents with up to eight channels on one side, and up to four on the other

Specifications
100 mL | |
Reservoir, 100mL, white | |
Non-sterile | |
10 | |
Single-use |
white | |
100 mL | |
Reagent Reservoir | |
Not Autoclavable |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2129113 | Certificate of Compliance | 06/02/2023 | 9510047 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.