Learn More
Thermo Scientific™ Fixed Needle Autosampler Syringes, 1000uL, for Thermo Scientific™ Instruments
Gas-tight syringe, headspace
Brand: Thermo Scientific™ 365Q2121
341.92 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
23 G | |
1 mL | |
Side Hole |
Gas Tight GC Autosampler Syringe, Fixed Needle | |
65 mm | |
TriPlus RSH Autosampler |
Product Suggestions
Customers who viewed this item also viewed
Special Offers
-
Multiple Manufacturers
Dairy and Plant-Based Products Testing Promotion
Quality testing products for all your needs.Promo Code: 24134Expires: 2025-03-31