missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SIGNAL Blood Culture System

Detect the growth of aerobic, anaerobic and micro-aeophilic organisms from blood samples with this unique one bottle blood culture system. Incubator trays (BC0104A) can be ordered separately, where required.

Brand:  Thermo Scientific™ BC0100M

326.51 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 11933192

  • £637.00 / Pack of 20

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Detect the growth of aerobic, anaerobic and micro-aerophilic organisms from blood samples using Thermo Scientific™ Oxoid™ SIGNAL Blood Culture System. SIGNAL Blood Culture System is a unique one bottle manual media system accommodating all patient populations and scenarios. The system uses a special broth medium which enables a wide range of aerobes, anaerobes and microaerophilic organisms to be cultured in a single bottle. A positive result is indicated when the blood/broth mixture is forced into the signal device by the pressure created by growing organisms.

The blood system is normally a sterile environment, so the detection of bacteria in blood (bacteraemia) is always abnormal. Early detection of bacteraemia is of utmost importance because of the high mortality rates associated with this condition.

The SIGNAL Blood Culture System is a unique one-bottle system suitable for use with both adult and pediatric samples, and requires only the most basic laboratory equipment, making it suitable for high- or low-level-usage laboratories. The SIGNAL Blood Culture System is manufactured to the International Quality Standard ISO 9001:2000 and is CE marked according to the In Vitro Diagnostic Directive 98/79/EC.

  • Easy-to-read-results: the SIGNAL Blood Culture System detects gases produced during organism metabolism, which causes an increase in pressure in the sealed bottle, forcing some of the blood/broth solution into the SIGNAL growth indicator chamber device giving a clear, visual positive display.
  • Reduced contamination: removable lid on indicator device permits samples to be taken directly from the sampling chamber, rather than bottle broth reducing the risk of potential contamination.
  • Cost-effective: does not require sophisticated laboratory equipment, just one bottle is needed to detect most aerobes, anaerobes and microaerophilic organisms
  • Additional capabilities: although it is primarily designed for the culture of organisms from human blood samples, SIGNAL Blood Culture System is also effective at recovering organisms from other body fluids including joint, abdominal and CAPD (Continuous Peritoneal Dialysis). It is also shown to be effective for veterinary use.

SIGNAL Blood Culture System comes complete with blood culture bottles filled with the blood culture medium and growth indicator devices.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

SIGNAL Blood Culture System
Aerobic, anaerobic and micro-aerophilic organisms from blood samples
A sealed blood culture bottle containing 80mL of broth medium, a sterile growth indicator devices which is vented through a 0.2 micron hydrophobic membrane and an instruction leaflet.
20 Tests/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6251121 Certificate TypeCertificate of Analysis Date03/04/2025 Catalog NumberBC0100M
Lot Number6251123 Certificate TypeCertificate of Analysis Date03/04/2025 Catalog NumberBC0100M
Lot Number6251122 Certificate TypeCertificate of Analysis Date03/04/2025 Catalog NumberBC0100M
Lot Number6244631 Certificate TypeCertificate of Analysis Date28/03/2025 Catalog NumberBC0100M
Lot Number6244630 Certificate TypeCertificate of Analysis Date28/03/2025 Catalog NumberBC0100M
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ SIGNAL Blood Culture System > Blood Culture System

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.