Learn More
Thermo Scientific™ Workstation/Stacking System for Tubes/Plates
Convenient platform for tube handling and reagent dispensing .
Brand: Thermo Scientific™ AB0538
43.58 GBP valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Description
Highlights
- Convenient platform for tube handling and reagent dispensing
- Holds:
- 96 x 0.2ml V-bottom tubes
- 96 x 0.3ml U-bottom Omnistrips
- Thermo-Tube Plates
- Thermo-Fast 96 Non-Skirted Plate
- Thermo-Fast 96 Semi-Skirted Plate
- 48 x 0.5ml Thermo-Tubes - Multiple workstations can be securely stacked
- Can be stored at -190°C
Hazardous: No
Storage Condition: Ambient

Specifications
Real-Time PCR Systems or Thermal Cyclers | |
10 units |
Product Suggestions
Customers who viewed this item also viewed
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Days
29Hours
7Minutes
7Seconds
56
Your input is important to us. Please complete this form to provide feedback related to the content on this product.