Learn More
Cole-Parmer™ Digi-Sense™ Insulated-Wire Thermocouple
Use these probes for a variety of high-temperature applications. Cole-Parmer™ Digi-Sense™ Insulated-Wire Thermocouple Probes include straight insulated cable with no handle and feature ANSI color-coded mini connectors: type J - black, type K - yellow, and type T - blue.
£44.70 - £121.00
Code produit | Marque | Length (English) | Prix | Quantité et disponibilité | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Length (English) | Prix | Quantité et disponibilité | ||||||
11841280
|
Cole-Parmer™
08512-82 |
120 in. (probe) |
£44.70
Each |
Estimated Shipment: 10-06-2025
Log in to see stock. |
||||||
11706624
|
Cole-Parmer™
08505-85 |
60 in. (probe) |
£121.00
Pack of 6 |
Estimated Shipment: 13-06-2025
Log in to see stock. |
||||||
Description
- Flexible Kapton-insulated wire is ideal for checking food temperatures
- Flexible fiberglass-Insulated wire used for high-temperature measurements
- Custom designs or changes to this product are available upon request
- Exposed-junction probes are not recommended for use with aqueous solutions
- Do not subject type J exposed-junction probes to moisture as they will rust
Spécification
Thermocouple Probe |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Cole-Parmer™ Digi-Sense™ Insulated-Wire Thermocouple