missing translation for 'onlineSavingsMsg'
Learn More
Thieno[3,2-b]thiophene, 97%, Thermo Scientific Chemicals Product Code.: 15483967

Thieno[3,2-b]thiophene, 97%, Thermo Scientific Chemicals

Product Code. 15483967
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15483967

Brand: Thermo Scientific Alfa Aesar H30637.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 251-41-2
Molecular Formula C6H4S2
Molecular Weight (g/mol) 140.218
MDL Number MFCD00179479
InChI Key VJYJJHQEVLEOFL-UHFFFAOYSA-N
Synonym thieno 3,2-b thiophene, 1,4-dithiapentalene, thienothiophene, 1,4-thiophthene, pubchem21292, acmc-209ghi, thieno 3,2-b thiophen, thieno 3,2-b-thiopheneShow MoreShow Less
PubChem CID 136063
IUPAC Name thieno[3,2-b]thiophene
SMILES C1=CSC2=C1SC=C2

Specifications

Melting Point 54°C to 58°C
Boiling Point 71°C (1 mmHg)
Quantity 5 g
Formula Weight 140.23
Percent Purity 97%
Chemical Name or Material Thieno[3,2-b]thiophene

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10210327 Certificate TypeCertificate of Analysis Date25/05/2023 Catalog NumberH30637.06
Lot Number5002P12Q Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH30637.03,H30637.06
Lot Number10149551 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH30637.03,H30637.06
Lot Number5003F30W Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH30637.03,H30637.06
Lot Number5002L15W Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH30637.03,H30637.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thieno[3,2-b]thiophene, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.