missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Thrombin Protease
For site-specific separation of the GST tag from proteins expressed using pGEX-T vectors
Brand: Cytiva 27-0846-01
130.15 GBP valid until 2025-02-21
Use promo code "24115" to get your promotional price.
This item is not returnable.
View return policy
Description
- Supplied as lyophilized powder
- Enables the site-specific cleavage of fusion proteins containing an accessible thrombin recognition sequence
- Purified from bovine plasma; functionally free of other clotting factors, plasminogen, and plasmin
- Can be used either following affinity purification or while fusion proteins are bound to Glutathione Sepharose™ 4B, Glutathione Sepharose High Performance, or Glutathione Sepharose 4 Fast Flow
Specifications
Elution Buffer: 50mM Tris HCl, 10mM reduced Glutathione pH 8, Cleavage Buffer: PBS: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4 pH 7.3 | |
Thrombin Protease Fusion Protein | |
500U | |
Site-specific separation of the GST tag from proteins expressed using pGEX-T vectors, one unit of enzyme cleaves 90% of 100μg of a test GST fusion protein when incubated in 1X PBS at 22°C for 16 hours |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : thrombin
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytiva Thrombin Protease > 500 units
Spot an opportunity for improvement?Share a Content Correction