Learn More
Thulium(III) acetate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Description
Thulium Acetate is the important dopant for silica-based fiber amplifiers. It can also be used in portable X-ray devices that has been bombarded in a nuclear reactor as a radiation source. Used as laboratory reagent, optical glasses, structural ceramics, catalysts, electrical components and photo-optical material. The other main application is used for optical fiber.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Physical Form | Crystalline Aggregates |
Quantity | 10 g |
Solubility Information | Moderately soluble in water,strong mineral acids. |
Formula Weight | 346.07 (Anhydrous) |
Percent Purity | 99.9% |
Grade | Reacton™ |
Sensitivity | Hygroscopic |
Assay | (REO) |
Chemical Name or Material | Thulium(III) acetate hydrate |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
P09J015 | Certificate of Analysis | 09/05/2023 | 014582.09,014582.04 |
N28J044 | Certificate of Analysis | 09/05/2023 | 014582.09,014582.04 |
N17I015 | Certificate of Analysis | 12/04/2022 | 014582.09,014582.04 |
M20I025 | Certificate of Analysis | 12/04/2022 | 014582.09,014582.04 |
N24H004 | Certificate of Analysis | 29/11/2021 | 014582.09,014582.04 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.