missing translation for 'onlineSavingsMsg'
Learn More
Thulium(III) acetate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11312538

Thulium(III) acetate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11312538
2 g
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy

Product Code. 11312538

Brand: Thermo Scientific Alfa Aesar 014582.04

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thulium Acetate is the important dopant for silica-based fiber amplifiers. It can also be used in portable X-ray devices that has been bombarded in a nuclear reactor as a radiation source. Used as laboratory reagent, optical glasses, structural ceramics, catalysts, electrical components and photo-optical material. The other main application is used for optical fiber.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 314041-04-8
Molecular Formula C6H9O6Tm
Molecular Weight (g/mol) 346.07
MDL Number MFCD00013046
InChI Key SNMVVAHJCCXTQR-UHFFFAOYSA-K
Synonym acmc-1afih, thulium acetate-water 1/3/1, thulium 3+ ion triacetate hydrate
IUPAC Name thulium(3+) triacetate
SMILES [Tm+3].CC([O-])=O.CC([O-])=O.CC([O-])=O

Specifications

Physical Form Crystalline Aggregates
Quantity 2 g
Solubility Information Moderately soluble in water,strong mineral acids.
Formula Weight 346.07 (Anhydrous)
Percent Purity 99.9%
Grade Reacton™
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Thulium(III) acetate hydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP09J015 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog Number014582.09,014582.04
Lot NumberN28J044 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog Number014582.09,014582.04
Lot NumberN17I015 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number014582.09,014582.04
Lot NumberM20I025 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number014582.09,014582.04
Lot NumberN24H004 Certificate TypeCertificate of Analysis Date29/11/2021 Catalog Number014582.09,014582.04

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thulium(III) acetate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.