missing translation for 'onlineSavingsMsg'
Learn More
Tiglic acid, 97+%, Thermo Scientific Chemicals Product Code.: 11441618

Tiglic acid, 97+%, Thermo Scientific Chemicals

Product Code. 11441618
500 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11441618

Brand: Thermo Scientific Alfa Aesar A13905.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-59-1
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00066864
InChI Key UIERETOOQGIECD-ONEGZZNKSA-N
Synonym tiglic acid, tiglinic acid, cevadic acid, 2-methyl-2-butenoic acid, trans-2-methylcrotonic acid, trans-2,3-dimethylacrylic acid, 2-methylbut-2-enoic acid, e-2-methyl-2-butenoic acid, trans-2-methyl-2-butenoic acid, e-2,3-dimethylacrylic acidShow MoreShow Less
PubChem CID 125468
ChEBI CHEBI:9592
IUPAC Name (E)-2-methylbut-2-enoic acid
SMILES C\C=C(/C)C(O)=O

Specifications

Melting Point 62°C to 66°C
Boiling Point 198°C to 199°C
UN Number UN3261
Beilstein 1236500
Merck Index 149433
Formula Weight 100.12
Percent Purity ≥98%
Density 0.969
Quantity 500 g
Flash Point 101°C (213°F)
Chemical Name or Material Tiglic acid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10252968 Certificate TypeCertificate of Analysis Date05/11/2024 Catalog NumberA13905.22,A13905.14,A13905.36
Lot Number10248959 Certificate TypeCertificate of Analysis Date18/12/2023 Catalog NumberA13905.22,A13905.14,A13905.36
Lot Number10241222 Certificate TypeCertificate of Analysis Date22/07/2022 Catalog NumberA13905.22,A13905.14,A13905.36
Lot Number10139192 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13905.22,A13905.14,A13905.36
Lot Number10165905 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13905.22,A13905.14,A13905.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tiglic acid, 97+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.