missing translation for 'onlineSavingsMsg'
Learn More
Tin(IV) oxide, 99.9% (metals basis), Thermo Scientific Chemicals Product Code.: 11321629

Tin(IV) oxide, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11321629
500 g
Click to view available options
Quantity:
100 g
500 g
2 kg
This item is not returnable. View return policy

Product Code. 11321629

Brand: Thermo Scientific Alfa Aesar 012283.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is employed in the manufacture of white enamels and for polishing glass. It has a broad range of applications in many areas such as semiconductors, photovoltaic materials, solar cells, lithium ion batteries, photocatalysts, and as gas-sensing agents in solid-state detectors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18282-10-5
Molecular Formula O2Sn
Molecular Weight (g/mol) 150.71
MDL Number MFCD00011244
InChI Key XOLBLPGZBRYERU-UHFFFAOYSA-N
Synonym tin dioxide, tin iv oxide, stannic oxide, cassiterite, flowers of tin, stannic dioxide, tin oxide sno2, tin peroxide, white tin oxide, stannic anhydrideShow MoreShow Less
PubChem CID 29011
ChEBI CHEBI:52991
SMILES O=[Sn]=O

Specifications

Boiling Point 1800°C to 1900°C (sublimation)
Melting Point 1630°C
Color White to Gray
Physical Form Powder
Quantity 500 g
Assay Percent Range 99.9% metal basis
Solubility Information Soluble in hot concentrated alkalis,and concentrated acids. Insoluble in alcohol.
IUPAC Name stannanedione
Formula Weight 150.69
Odor Odorless
Refractive Index 1.9968
Density 6.95
Chemical Name or Material Tin(IV) oxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP05L041 Certificate TypeCertificate of Analysis Date07/03/2025 Catalog Number012283.36,012283.A3,012283.22
Lot NumberS10K010 Certificate TypeCertificate of Analysis Date20/06/2024 Catalog Number012283.36,012283.A3,012283.22,012283.A1
Lot NumberQ14I033 Certificate TypeCertificate of Analysis Date11/10/2023 Catalog Number012283.36,012283.A3,012283.22,012283.A1
Lot NumberT28J039 Certificate TypeCertificate of Analysis Date07/08/2023 Catalog Number012283.36,012283.A3,012283.22,012283.A1
Lot NumberX28C003 Certificate TypeCertificate of Analysis Date09/02/2023 Catalog Number012283.36,012283.A3,012283.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tin(IV) oxide, 99.9% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.