Learn More
Titanium(IV) chloride, 99.0% min, typically 99.6%, Thermo Scientific Chemicals
For Use in Manufacture of Titanium Pigments and Metal, As a Catalyst in organic Syntheses, in Iridescent Glass
£28.60 - £224.00
Chemical Identifiers
CAS | 7550-45-0 |
---|---|
Molecular Formula | Cl4Ti |
Molecular Weight (g/mol) | 189.667 |
MDL Number | MFCD00011267 |
InChI Key | XJDNKRIXUMDJCW-UHFFFAOYSA-J |
Synonym | titanium iv chloride, titanium 4+ tetrachloride, acmc-1bjgv, titanium 4+ ion tetrachloride, parent, titanium +4 cation tetrachloride, titanium iv chloride 100ml, superlist names titanium chloride, t-4, titanic chloride; titanio tetracloruro di, unii-8o3pje5t7q; titanium chloride ticl4Show More |
PubChem CID | 160960 |
IUPAC Name | titanium(4+);tetrachloride |
SMILES | [Cl-].[Cl-].[Cl-].[Cl-].[Ti+4] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15401077
|
Thermo Scientific Alfa Aesar
014713.AE |
100 mL | N/A | ||||||
15411077
|
Thermo Scientific Alfa Aesar
014713.K2 |
1 L | N/A | ||||||
15421077
|
Thermo Scientific Alfa Aesar
014713.L7 |
3 x 1 L | N/A | ||||||
Description
In manufacture of titanium pigments and metal, as a catalyst in organic syntheses, in iridescent glass, as a smoke screenTitanium(IV) chloride is used to make titanium metal, pigment titanium dioxide and is used in smoke screens. It is involved in the activation of pyrrolidines for better conversion through a modified Bouveault reaction, to the corresponding alpha, alpha -dimethylamines. In organometllic chemistry, it is used to prepare complexes like titanocene dichloride, which is a precursor to Ziegler-Natta catalysts. Tebbe’s reagent is an aluminum containing derivative of titanocene used for the olefination reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7550-45-0 | |
189.667 | |
XJDNKRIXUMDJCW-UHFFFAOYSA-J | |
160960 | |
[Cl-].[Cl-].[Cl-].[Cl-].[Ti+4] |
Cl4Ti | |
MFCD00011267 | |
titanium iv chloride, titanium 4+ tetrachloride, acmc-1bjgv, titanium 4+ ion tetrachloride, parent, titanium +4 cation tetrachloride, titanium iv chloride 100ml, superlist names titanium chloride, t-4, titanic chloride; titanio tetracloruro di, unii-8o3pje5t7q; titanium chloride ticl4Show More | |
titanium(4+);tetrachloride |
Specifications
136°C to 137°C | |
7550-45-0 | |
Liquid | |
Cl4Ti | |
titanium iv chloride, titanium 4+ tetrachloride, acmc-1bjgv, titanium 4+ ion tetrachloride, parent, titanium +4 cation tetrachloride, titanium iv chloride 100ml, superlist names titanium chloride, t-4, titanic chloride; titanio tetracloruro di, unii-8o3pje5t7q; titanium chloride ticl4 | |
XJDNKRIXUMDJCW-UHFFFAOYSA-J | |
titanium(4+);tetrachloride | |
160960 | |
1.61 | |
Colorless to yellow liquid | |
Titanium(IV) chloride |
25°C | |
Yellow | |
99.0% min., typically 99.6% | |
MFCD00011267 | |
Soluble in dilute hydrochloric acid,alcohol and alkalies.Miscible with ethanol,hydrochloric acid and organic solvents. | |
[Cl-].[Cl-].[Cl-].[Cl-].[Ti+4] | |
189.667 | |
189.69 | |
Moisture sensitive | |
1.726 g/cm3 |
Safety and Handling
P260-P264b-P271-P280-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H330-H335
missing translation for 'dotInformation' : Hazard Class: 6.1, Packing Group: I
missing translation for 'einecsNumber' : 231-441-9
missing translation for 'rtecsNumber' : XR1925000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.