missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) ethoxide, TiO{2} 33% min, Thermo Scientific Chemicals

£23.72 - £78.50

Chemical Identifiers

CAS 3087-36-3
Molecular Formula C8H20O4Ti
Molecular Weight (g/mol) 228.111
MDL Number MFCD00009071
InChI Key JMXKSZRRTHPKDL-UHFFFAOYSA-N
Synonym titanium iv ethoxide, ethyl titanate, titanium tetraethoxide, titanium ethoxide, tetraethyl titanate, titanium iv ethanolate, titanium 4+ tetraethanolate, titanium 4+ ethanolate
PubChem CID 76524
IUPAC Name ethanolate;titanium(4+)
SMILES CC[O-].CC[O-].CC[O-].CC[O-].[Ti+4]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15492957
View Documents
Thermo Scientific Alfa Aesar
077142.06
5 g
£23.72
5g
Please to purchase this item. Need a web account? Register with us today!
15402967
View Documents
Thermo Scientific Alfa Aesar
077142.18
50 g
£30.25
50g
Please to purchase this item. Need a web account? Register with us today!
15412967
View Documents
Thermo Scientific Alfa Aesar
077142.30
250 g
£78.50
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Titanium(IV) ethoxide is used in organic synthesis and material science. It is involved in the synthesis of anti-perspirants and antifungal agents. Furthermore, it is used to prepare carbon films in association with silicon dioxide. It is also used as a precursor for the synthesis of Ti4+ ions titanium nanoparticles by various methods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3087-36-3
228.111
JMXKSZRRTHPKDL-UHFFFAOYSA-N
76524
CC[O-].CC[O-].CC[O-].CC[O-].[Ti+4]
C8H20O4Ti
MFCD00009071
titanium iv ethoxide, ethyl titanate, titanium tetraethoxide, titanium ethoxide, tetraethyl titanate, titanium iv ethanolate, titanium 4+ tetraethanolate, titanium 4+ ethanolate
ethanolate;titanium(4+)

Specifications

150°C to 152°C (10mmHg)
3087-36-3
Liquid
Ti(OCH2CH3)4
UN2924
titanium iv ethoxide, ethyl titanate, titanium tetraethoxide, titanium ethoxide, tetraethyl titanate, titanium iv ethanolate, titanium 4+ tetraethanolate, titanium 4+ ethanolate
JMXKSZRRTHPKDL-UHFFFAOYSA-N
ethanolate;titanium(4+)
76524
TiO2 33% min
1.5043
1.088 g/mL
-40°C
Yellow
C8H20O4Ti
MFCD00009071
28°C (84°F)
Soluble in organic solvents; Decomposes in water.
CC[O-].CC[O-].CC[O-].CC[O-].[Ti+4]
228.111
228.15
Alcohol-like
Moisture sensitive
Titanium (IV) ethoxide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H319
Flammable liquid and vapor.
Causes serious eye irritation.

P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P370+P378q-P501c

H226-H319-H336

missing translation for 'dotInformation' : Hazard Class: 3; Packaging Group: III

missing translation for 'einecsNumber' : 221-410-8

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Argon

Product Identifier
  • Titanium(IV) ethoxide, TiO2 33% min

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P240-Ground/bond container and receiving equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..

Supplemental information
  • MIXTURE LIST-Contains : Titanium(IV) ethoxide, Titanium(IV) isopropoxide

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only