Learn More
Titanium(IV) n-butoxide, 98+%, Thermo Scientific Chemicals
£91.30 - £1020.00
Chemical Identifiers
CAS | 5593-70-4 |
---|---|
Molecular Formula | C16H36O4Ti |
Molecular Weight (g/mol) | 340.35 |
MDL Number | MFCD00009433 |
InChI Key | FPCJKVGGYOAWIZ-UHFFFAOYSA-N |
Synonym | butyl titanate, butan-1-ol; titanium, titanium n-butoxide, tetrabutyl orthotitanate, tetrabutyl titanate, 1-butanol; titanium, tetra butyl orthotitanate, tetrabutyl ortho-titanate |
PubChem CID | 25203927 |
IUPAC Name | butan-1-ol;titanium |
SMILES | CCCCO.CCCCO.CCCCO.CCCCO.[Ti] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10216470
|
Thermo Scientific Acros
223195000 |
500 g | Glass bottle |
£91.30
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10104900
|
Thermo Scientific Acros
223190010 |
1 kg | Glass bottle |
£146.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10782261
|
Thermo Scientific Acros
223190020 |
2 kg | Glass bottle |
£282.00
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15204694
|
Thermo Scientific Acros
223190100 |
10 kg | Plastic Drum |
£1020.00
10kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acidChemical Identifiers
5593-70-4 | |
340.35 | |
FPCJKVGGYOAWIZ-UHFFFAOYSA-N | |
25203927 | |
CCCCO.CCCCO.CCCCO.CCCCO.[Ti] |
C16H36O4Ti | |
MFCD00009433 | |
butyl titanate, butan-1-ol; titanium, titanium n-butoxide, tetrabutyl orthotitanate, tetrabutyl titanate, 1-butanol; titanium, tetra butyl orthotitanate, tetrabutyl ortho-titanate | |
butan-1-ol;titanium |
Specifications
5593-70-4 | |
Colorless to Yellow | |
310.0°C to 314.0°C | |
Authentic | |
C16H36O4Ti | |
1.486 | |
0.99 | |
Solubility in water: rapidly hydrolized by water. Other solubilities: miscible with most common organic solvents | |
CCCCO.CCCCO.CCCCO.CCCCO.[Ti] | |
340.35 | |
50-80 mPa.s (20°C) | |
98+% | |
Titanium(IV) n-butoxide, 99% |
-75°C | |
0.9900g/mL | |
47°C | |
98+% | |
Ti[O(CH2)3CH3]4 | |
MFCD00009433 | |
butyl titanate, butan-1-ol; titanium, titanium n-butoxide, tetrabutyl orthotitanate, tetrabutyl titanate, 1-butanol; titanium, tetra butyl orthotitanate, tetrabutyl ortho-titanate | |
FPCJKVGGYOAWIZ-UHFFFAOYSA-N | |
butan-1-ol;titanium | |
25203927 | |
340.35 | |
Liquid |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
May cause drowsiness or dizziness.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
If skin irritation occurs: Get medical advice/at
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 227-006-8
missing translation for 'rtecsNumber' : XR1585000
missing translation for 'tsca' : TSCA
Recommended Storage : the product darkens with age
- Titanium(IV) n-butoxide
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
RUO – Research Use Only