Learn More
MedChemExpress TMB-PS
MedChemExpress TMB-PS(102062-36-2) is N-(3-sulfopropyl)-3,3',5,5'-tetramenthylbenzidine sodium salt; White - pale yellow crystalline powder, soluble in water, can be used under neutral, acidic and alkaline conditions.
Brand: MedChemExpress HY-15931

Specifications
1 g | |
DMSO : ≥ 35 mg/mL (96.55 mM) | |
102062-36-2 | |
TMBZ-PS |
Powder -20°C 3 years, 4°C 2 years ∣ In solvent -80°C 6 months, -20°C 1 month | |
Room temperature | |
362.49 g/mol |
Product Suggestions
Customers who viewed this item also viewed
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.