missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ ToughStripe™ Anti-Slip Floor Safety Sign
Hard-to-miss signs made of durable floor marking tape
£82.50 - £118.00
Specifications
Certifications/Compliance | ASTM ANSI F1679, ANSI A1264.2-2006, OSHA |
---|---|
Finish | Gloss |
Material | Polyester |
Quantity | 1 Ea. |
Adhesive Type | Permanent Rubber |
Product Code | Brand | Displayed Text | Color | Shape | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Displayed Text | Color | Shape | Price | Quantity & Availability | |||
11751705
|
Brady™
104492 |
Fire Extinguisher | Black on White | Round |
£118.00
Each |
Estimated Shipment: 06-09-2024
Log in to see stock. |
|||
11781705
|
Brady™
104495 |
Emergency Exit | Black on White | Round |
£118.00
Each |
Estimated Shipment: 06-09-2024
Log in to see stock. |
|||
11731705
|
Brady™
104490 |
Exit | Black on White | Round |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11741705
|
Brady™
104491 |
First Aid | White | Rectangular |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11771705
|
Brady™
104494 |
Fire Hose | Black on White | Round |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11701705
|
Brady™
104487 |
Safety Shower | Black | Round |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11791695
|
Brady™
104486 |
First Aid | White | Round |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11761705
|
Brady™
104493 |
Fire Alarm | Black on White | Round |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11711715
|
Brady™
104498 |
Chemical Storage | Red | Rectangular |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11721715
|
Brady™
104499 |
Watch Your Step | Black on Yellow | Rectangular |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11756355
|
Brady™
104502 |
Ear Protection Area | Black on Yellow | Rectangular |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11766355
|
Brady™
104503 |
Eye Protection Area | Black on Yellow | Rectangular |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11776355
|
Brady™
104504 |
Hard Hat Area | Black on Yellow | Rectangular |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11796355
|
Brady™
104506 |
BIOHAZARD | Black on Orange | Rectangular |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11786355
|
Brady™
104505 |
Safety Goggles Required | Black on Yellow | Rectangular |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11706365
|
Brady™
104507 |
Eye Wash Station | Black | Round |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11736365
|
Brady™
104510 |
Safety Glasses Required | Black on Yellow | Rectangular |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
- Backed with high-performance adhesive
- Features a highly visible, glossy appearance for communicating important safety and warning messages to employees
- Clear anti-skid area
- Designed with sections of anti-skid material that help prevent accidental slips, trips and falls
- Easy to remove and apply
- Made of polyester
Specifications
ASTM ANSI F1679, ANSI A1264.2-2006, OSHA | |
Polyester | |
Permanent Rubber | |
Used to mark aisles, passageways, storage locations, point out safety equipment and exits and enforce protective equipment requirements with glossy appearance and clear anti-skid area for communicating important safety and warning messages to employees | |
-0.4°F | |
-18°C to 54°C | |
English |
Gloss | |
1 Ea. | |
ToughStripe - Pre-printed Floor Signs | |
129.9°F | |
0.01 in. | |
-0.4°F to 129.9 °F | |
Floor Marking Safety Sign |