Learn More
Brady™ ToughStripe Max Solid Floor Tape
ToughStripe™ Max Floor Marking Tape is purpose-built to hold its ground in forklift and heavy traffic areas.
£129.62 - £185.98
Specifications
Adhesive Type | Rubber |
---|---|
Material | Polyester |
Product Type | Floor Tape |
Finish | Gloss |
Length (English) | 100 ft. |
Product Code | Brand | Color | Width (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Width (Metric) | Price | Quantity & Availability | |||||
30280394
|
Brady™
177014 |
Yellow | 76.2 mm |
£163.43
1 roll |
Estimated Shipment: 19-06-2025
Log in to see stock. |
|||||
30280374
|
Brady™
177015 |
Yellow | 50.8 mm |
£129.62
1 roll |
Estimated Shipment: 19-06-2025
Log in to see stock. |
|||||
30280344
|
Brady™
177016 |
Yellow | 101.6 mm |
£185.98
1 roll |
Estimated Shipment: 16-05-2025
Log in to see stock. |
|||||
30280361
|
Brady™
177019 |
Blue | 50.8 mm |
£129.62
1 roll |
Estimated Shipment: 19-06-2025
Log in to see stock. |
|||||
30280342
|
Brady™
177020 |
Red | 50.8 mm |
£129.62
1 roll |
Estimated Shipment: 16-05-2025
Log in to see stock. |
|||||
30280350
|
Brady™
177021 |
White | 50.8 mm |
£129.62
1 roll |
Estimated Shipment: 16-05-2025
Log in to see stock. |
|||||
30280379
|
Brady™
177022 |
Green | 50.8 mm |
£129.62
1 roll |
Estimated Shipment: 19-06-2025
Log in to see stock. |
|||||
30280349
|
Brady™
177023 |
Blue | 76.2 mm |
£163.43
1 roll |
Estimated Shipment: 19-06-2025
Log in to see stock. |
|||||
30280360
|
Brady™
177026 |
White | 76.2 mm |
£163.43
1 roll |
Estimated Shipment: 16-05-2025
Log in to see stock. |
|||||
30280372
|
Brady™
177027 |
White | 101.6 mm |
£185.98
1 roll |
Estimated Shipment: 16-05-2025
Log in to see stock. |
|||||
30280371
|
Brady™
177028 |
Red | 76.2 mm |
£163.43
1 roll |
Estimated Shipment: 16-05-2025
Log in to see stock. |
|||||
30280366
|
Brady™
177030 |
Orange | 50.8 mm |
£129.62
1 roll |
Estimated Shipment: 19-06-2025
Log in to see stock. |
|||||
30280359
|
Brady™
177031 |
Green | 76.2 mm |
£163.43
1 roll |
Estimated Shipment: 16-05-2025
Log in to see stock. |
|||||
30280391
|
Brady™
177032 |
Red | 101.6 mm |
£185.98
1 roll |
Estimated Shipment: 16-05-2025
Log in to see stock. |
|||||
30280367
|
Brady™
177034 |
Orange | 76.2 mm |
£163.43
1 roll |
Estimated Shipment: 16-05-2025
Log in to see stock. |
|||||
30280347
|
Brady™
177036 |
Green | 101.6 mm |
£185.98
1 roll |
Estimated Shipment: 19-06-2025
Log in to see stock. |
|||||
30280393
|
Brady™
177037 |
Blue | 101.6 mm |
£185.98
1 roll |
Estimated Shipment: 16-05-2025
Log in to see stock. |
|||||
30280382
|
Brady™
177038 |
Black | 50.8 mm |
£129.62
1 roll |
Estimated Shipment: 19-06-2025
Log in to see stock. |
|||||
30280341
|
Brady™
177042 |
Orange | 101.6 mm |
£185.98
1 roll |
Estimated Shipment: 19-06-2025
Log in to see stock. |
|||||
30280364
|
Brady™
177043 |
Purple | 76.2 mm |
£163.43
1 roll |
Estimated Shipment: 19-06-2025
Log in to see stock. |
|||||
30280378
|
Brady™
177045 |
Purple | 50.8 mm |
£129.62
1 roll |
Estimated Shipment: 19-06-2025
Log in to see stock. |
|||||
30280384
|
Brady™
177049 |
Black | 76.2 mm |
£163.43
1 roll |
Estimated Shipment: 19-06-2025
Log in to see stock. |
|||||
30280346
|
Brady™
177059 |
Black | 101.6 mm |
£185.98
1 roll |
Estimated Shipment: 19-06-2025
Log in to see stock. |
|||||
30280354
|
Brady™
177062 |
Gray | 50.8 mm |
£129.62
1 roll |
Estimated Shipment: 19-06-2025
Log in to see stock. |
|||||
30280375
|
Brady™
177068 |
Brown | 50.8 mm |
£129.62
1 roll |
Estimated Shipment: 19-06-2025
Log in to see stock. |
|||||
30280373
|
Brady™
177069 |
Brown | 101.6 mm |
£185.98
1 roll |
Estimated Shipment: 19-06-2025
Log in to see stock. |
|||||
30280348
|
Brady™
177070 |
Brown | 76.2 mm |
£163.43
1 roll |
Estimated Shipment: 19-06-2025
Log in to see stock. |
|||||
30280385
|
Brady™
177074 |
Purple | 101.6 mm |
£185.98
1 roll |
Estimated Shipment: 19-06-2025
Log in to see stock. |
|||||
30280368
|
Brady™
177078 |
Gray | 76.2 mm |
£163.43
1 roll |
Estimated Shipment: 19-06-2025
Log in to see stock. |
|||||
30280376
|
Brady™
177096 |
Gray | 101.6 mm |
£185.98
1 roll |
Estimated Shipment: 19-06-2025
Log in to see stock. |
|||||
Description
- Re-engineered to be twice as thin yet twice as durable to better withstand damaging impacts in forklift and heavy traffic areas
- Thinner profile and true edge-to-edge adhesive minimise pick points
- Strong polyester (PETG) material is four times more rigid than vinyl and contains no plasticisers
- Damage-isolating design keeps tape intact longer by preventing rips, gouges and tears from running
- Matt finish provides excellent colour contrast for clear visibility under any lighting
- Compact construction results in a smaller, lightweight roll
- Can be easily installed by one person with minimal tools
- Compatible with the ToughStripe™ Floor Tape Applicator (part number 150852)
Specifications
Rubber | |
Floor Tape | |
100 ft. | |
50°F to 140°F |
Polyester | |
Gloss | |
30.48 m | |
10°C to 60°C |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Brady™ ToughStripe Max Solid Floor Tape