Learn More
Fisherbrand™ Traceable™ pH/ORP Meter
Designed for water applications, this meter takes the guesswork out of measurements. Fisherbrand™ Traceable™ pH/ORP Meters is ideal for testing pH and ORP in wastewater, water quality and environmental applications.
Brand:  Fisherbrand™ 98769-31
Product Code. 15577583
  • £446.73 / Each
Explore more special offers
Description
Description
  • Compact instrument fits in the palm of your hand
  • Accuracy: ±0.02 pH
  • 25 point memory storage  with time/date stamp
  • Waterproof (IP67)
  • Triple Display shows pH or ORP measurement, temperature and time\date
  • Ideal for wastewater, water quality and environmental applications
Specifications
Specifications
±20mV
User-selected 1, 2 or 3-point calibration
4277 Traceable™ pH\ORP Meter
pH/ORP Meter
±1°C
0.1º (1º when >100º)
26 x 6.4 x 2.5cm
±0.02
-1800 to 1800 mV
2x CR2032 3V Batteries (included)
0.1mV <1000mV (1mV ≥1000mV)
0 to 60ºC
0 to 60ºC or 32 to 140ºF (selectable)
127 g
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Traceable™ pH/ORP Meter > pH/ORP Meter