missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TraceGOLD TG-5HT GC Columns

Thermo Scientific™ TraceGOLD TG-5HT GC Columns offer extended operation up to 400°C, ideal for high temperature GC applications.

£412.05 - £778.88

Specifications

Product Line TraceGOLD
Stationary Phase TraceGOLD™ TG-5 HT
USP Type G27, G36
Max. Temperature 380/400°C
For Use With (Application) Pesticides Analysis, GC/MS for Pesticides, GC/MS for Residual Solvents, Solvents, and Solvent Impurities
View More Specs

Products 9
Product Code Brand Length (Metric) Diameter (Metric) Inner Film Thickness Price Quantity & Availability  
Product Code Brand Length (Metric) Diameter (Metric) Inner Film Thickness Price Quantity & Availability  
10137714
View Documents
Thermo Scientific™
26095-0350
15 m 0.25 mm 0.1 μm
£412.05
Each
Estimated Shipment: 07-05-2025
to see stock.
Add to basket
 
13466846
View Documents
Thermo Scientific™
26095-1300
15 m 0.25 mm 0.25 μm
£496.47
Each
In Stock
to see stock.
Add to basket
 
10669105
View Documents
Thermo Scientific™
26095-0360
15 m 0.32 mm 0.1 μm
£451.25
Each
Estimated Shipment: 12-05-2025
to see stock.
Add to basket
 
10200345
View Documents
Thermo Scientific™
26095-0470
30 m 0.25 mm 0.1 μm
£663.30
Each
Estimated Shipment: 27-05-2025
to see stock.
Add to basket
 
10545065
View Documents
Thermo Scientific™
26095-0480
30 m 0.32 mm 0.1 μm
£748.73
Each
Estimated Shipment: 15-05-2025
to see stock.
Add to basket
 
10230195
View Documents
Thermo Scientific™
26095-0620
30 m 0.53 mm 0.15 μm
£778.88
Each
Estimated Shipment: 06-05-2025
to see stock.
Add to basket
 
10240195
View Documents Promotion Details
Thermo Scientific™
26095-1420
30 m 0.25 mm 0.25 μm
£689.83
Each
Estimated Shipment: 29-04-2025
to see stock.
Add to basket
 
10670636
View Documents
Thermo Scientific™
26095-1430
30 m 0.32 mm 0.25 μm
£713.55
Each
Estimated Shipment: 14-05-2025
to see stock.
Add to basket
 
13476846
View Documents
Thermo Scientific™
26095-1640
10 m 0.53 mm 0.15 μm
£437.18
Each
In Stock
to see stock.
Add to basket
 
Description

Description

Thermo Scientific™ TraceGOLD TG-5HT GC Columns offer extended operation up to 400°C, ideal for high temperature GC applications.

  • Low polarity, 5% diphenyl/95% dimethyl polysiloxane
  • Lower bleed and better inertness than comparable high-temperature columns
  • Special design of fused silica tubing extends column lifetime by up to 40%
  • Similar to Rxi™-5HT, BD-5HT, VF-5HT, ZB-5HT

Recommended for:

  • Phenols
  • Residual solvents
  • Drugs of abuse
  • Semivolatiles
  • Pesticides
  • PCBs
  • Solvent impurities
Specifications

Specifications

TraceGOLD
G27, G36
Pesticides Analysis, GC/MS for Pesticides, GC/MS for Residual Solvents, Solvents, and Solvent Impurities
TraceGOLD™ TG-5 HT
380/400°C
Low-Polar
Videos
SDS
Documents

Documents

Product Certifications

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ TraceGOLD TG-5HT GC Columns

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.