Learn More
Fisherbrand™ Translucent Silicone Stoppers
Prevents liquid chemical leaks or escape from container.
£7.75 - £88.00
Product Code | Brand | Diameter (Metric) Bottom | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Bottom | Price | Quantity & Availability | |||||
11507662
|
Fisherbrand™
|
10 mm |
£23.10
Pack of 20 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
11517662
|
Fisherbrand™
|
11 mm |
£14.29
Pack of 10 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
11537662
|
Fisherbrand™
|
15 mm |
£27.70
Pack of 10 |
Estimated Shipment: 28-08-2024
Log in to see stock. |
|||||
11557662
|
Fisherbrand™
|
19 mm |
£12.85
Pack of 5 |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||||
11567662
|
Fisherbrand™
|
21 mm |
£27.40
Pack of 5 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
11587662
|
Fisherbrand™
|
25 mm |
£7.75
Pack of 2 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
11577662
|
Fisherbrand™
|
23 mm |
£34.00
Pack of 5 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
11597662
|
Fisherbrand™
|
27 mm |
£20.50
Pack of 2 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
11507672
|
Fisherbrand™
|
29 mm |
£11.80
Pack of 2 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
11527672
|
Fisherbrand™
|
33 mm |
£18.80
Pack of 2 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
11537672
|
Fisherbrand™
|
35 mm |
£11.10
Each |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
11547672
|
Fisherbrand™
|
37 mm |
£12.00
Each |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
11557672
|
Fisherbrand™
|
39 mm |
£13.45
Each |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
11567672
|
Fisherbrand™
|
41 mm |
£16.45
Each |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
11587672
|
Fisherbrand™
|
45 mm |
£63.40
Each |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
11547652
|
Fisherbrand™
|
4 mm |
£13.35
Pack of 50 |
Estimated Shipment: 08-08-2024
Log in to see stock. |
|||||
11557652
|
Fisherbrand™
|
5 mm |
£11.90
Pack of 50 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
11517682
|
Fisherbrand™
|
57 mm |
£88.00
Each |
Estimated Shipment: 30-08-2024
Log in to see stock. |
|||||
11527682
|
Fisherbrand™
|
62 mm |
£43.90
Each |
Estimated Shipment: 23-08-2024
Log in to see stock. |
|||||
11537682
|
Fisherbrand™
|
67 mm |
£72.85
Each |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
11547682
|
Fisherbrand™
|
71 mm |
£69.90
Each |
Estimated Shipment: 27-08-2024
Log in to see stock. |
|||||
11577652
|
Fisherbrand™
|
7 mm |
£15.40
Pack of 50 |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||||
11597652
|
Fisherbrand™
|
9 mm |
£43.35
Pack of 20 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
Description
- Non-toxic, translucent
- Temperature range -60°C to 200°C
- High electrical resistance
- High resistance to many chemicals including ozone
Specifications
Translucent | |
Silicone | |
Silicone Stopper |