Learn More
Deutsch Neumann™ Transparent PVC Tubing
Excellent option for viewing the flow of liquids. Deutsch Neumann™ Transparent PVC Tubing is strong but lightweight and suitable for general laboratory use.
Brand: Deutsch Neumann™ 3501216
Description
- Transparent color
- Heat resistant to 60°C
- High flexibility and chemical resistance
- Excelllent resistance to abrasion, smooth inside and out
Specifications
ISO 9001:2008 | |
PVC | |
50 m | |
1 Each |
12 mm | |
16 mm | |
2 mm | |
Transparent |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.