missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Deutsch Neumann™ Transparent PVC Tubing
Excellent option for viewing the flow of liquids. Deutsch Neumann™ Transparent PVC Tubing is strong but lightweight and suitable for general laboratory use.
Brand: Deutsch Neumann™ 3511927
This item is not returnable.
View return policy
Description
- Transparent color
- Heat resistant to 60°C
- High flexibility and chemical resistance
- Excelllent resistance to abrasion, smooth inside and out
Specifications
ISO 9001:2008 | |
PVC | |
50 m | |
1 Each | |
12 bar at 20°C |
19 mm | |
27 mm | |
4 mm | |
Transparent |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Deutsch Neumann™ Transparent PVC Tubing > Inner Diameter: 19mm; Outer Diameter: 27mm
Spot an opportunity for improvement?Share a Content Correction