missing translation for 'onlineSavingsMsg'
Learn More
Tri-n-butyltin hydride, 97%, Thermo Scientific Chemicals Product Code.: 10615862

Tri-n-butyltin hydride, 97%, Thermo Scientific Chemicals

Product Code. 10615862
10 g, Glass bottle
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10615862

Brand: Thermo Scientific Acros 215730100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 688-73-3
Molecular Formula C12H28Sn
Molecular Weight (g/mol) 291.04
MDL Number MFCD00009416
InChI Key PIILXFBHQILWPS-UHFFFAOYSA-N
Synonym tributylstannanyl, tributlytin, tributylstannic hydride, tributyl tin, tri-n-butylstannane hydride, unii-4xdx163p3d, tin, tri-n-butyl-, hydride, tri-n-butylstannyl radical, stannane, tri-n-butyl-, hydride, tributyl-tinShow MoreShow Less
PubChem CID 3032732
IUPAC Name tributyltin
SMILES CCCC[Sn](CCCC)CCCC

Specifications

Color Colorless
Density 1.0800g/mL
Boiling Point 80.0°C (0.4 mmHg)
Flash Point 40°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula [CH3(CH2)3]3SnH
Refractive Index 1.4720 to 1.4740
Quantity 10 g
Fieser 01,1192; 02,424; 03,294; 04,518; 05,685; 06,604; 07,379; 08,497; 09,476; 10,411; 11,545; 12,516; 14,312; 15,325; 16,343; 17,351
Merck Index 15, 9783
Specific Gravity 1.08
Solubility Information Solubility in water: reacts. Other solubilities: soluble in most organic solvents
Viscosity 1.5 mPa.s (20°C)
Formula Weight 291.04
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Tri-n-butyltin hydride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0461230 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number215730100,215730500,215732500
Lot NumberA0454383 Certificate TypeCertificate of Analysis Date23/04/2024 Catalog Number215730100,215730500,215732500
Lot NumberA0449260 Certificate TypeCertificate of Analysis Date26/03/2024 Catalog Number215730100,215730500,215732500
Lot NumberA0461230 Certificate TypeCertificate of Analysis Date20/03/2024 Catalog Number215730100,215730500,215732500
Lot NumberA0395042 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number215730100,215730500,215732500
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tri-n-butyltin hydride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H360FD-May damage fertility. May damage the unborn child.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H312-Harmful in contact with skin.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tri-n-butyltin hydride, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.