missing translation for 'onlineSavingsMsg'
Learn More
Trichloroacetyl chloride, 99%, Thermo Scientific Chemicals Product Code.: 11460257

Trichloroacetyl chloride, 99%, Thermo Scientific Chemicals

Product Code. 11460257
250 g
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy

Product Code. 11460257

Brand: Thermo Scientific Alfa Aesar B21675.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Trichloroacetyl chloride is used in the preparation of dihydro-1H-benzindoles. It is also used in the synthesis of 3-alkylbenzoxazolones It is used in the manufacture of pharmaceuticals and plant protection compounds. It is used for manufacturing the esters and anhydrides of trichloroacetic acid. It is used in the manufacture of pharmaceuticals and organic synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trichloroacetyl chloride is used in the preparation of dihydro-1H-benzindoles. It is also used in the synthesis of 3-alkylbenzoxazolones It is used in the manufacture of pharmaceuticals and plant protection compounds. It is used for manufacturing the esters and anhydrides of trichloroacetic acid. It is used in the manufacture of pharmaceuticals and organic synthesis intermediate.

Solubility
Reacts violently with water.

Notes
Moisture sensitive. Store away from oxidizing agents, bases, alcohols, amines and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-02-8
Molecular Formula C2Cl4O
Molecular Weight (g/mol) 181.821
MDL Number MFCD00000792
InChI Key PVFOMCVHYWHZJE-UHFFFAOYSA-N
Synonym trichloroacetyl chloride, superpalite, acetyl chloride, trichloro, trichloroacetochloride, trichloroacetic acid chloride, ccl3cocl, trichloroacetylchloride, unii-9sn86t76y6, ccris 6764, acetyl chloride, 2,2,2-trichloroShow MoreShow Less
PubChem CID 6420
IUPAC Name 2,2,2-trichloroacetyl chloride
SMILES C(=O)(C(Cl)(Cl)Cl)Cl

Specifications

Melting Point -146°C
Density 1.629
Boiling Point 114°C to 116°C
Refractive Index 1.47
Quantity 250 g
UN Number UN2442
Beilstein 774120
Sensitivity Moisture sensitive
Solubility Information Reacts violently with water.
Formula Weight 181.83
Percent Purity 99%
Chemical Name or Material Trichloroacetyl chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN18L066 Certificate TypeCertificate of Analysis Date20/02/2025 Catalog NumberB21675.18,B21675.0B,B21675.30
Lot NumberW04K032 Certificate TypeCertificate of Analysis Date11/09/2024 Catalog NumberB21675.18,B21675.0B,B21675.30
Lot NumberP26K001 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog NumberB21675.18,B21675.0B,B21675.30
Lot Number10249503 Certificate TypeCertificate of Analysis Date31/01/2024 Catalog NumberB21675.18,B21675.0B,B21675.30
Lot Number10215860 Certificate TypeCertificate of Analysis Date18/01/2024 Catalog NumberB21675.18,B21675.0B,B21675.30

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Trichloroacetyl chloride, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.